7-Chloro-4-nitro-2-phenylindole

Supplier: Matrix Scientific

024898-250MG
101819-172EA 303.08 USD
101819-172
7-Chloro-4-nitro-2-phenylindole
7-Chloro-4-nitro-2-phenylindole

Hazard: Irritant

Formula: C₁₄H₉ClN₂O₂
MDL Number: MFCD09743722
CAS Number: 663177-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR