1,3-Dimethyl-5-iodopyrazole

Supplier: Matrix Scientific

024896-500MG
101819-168EA 240 USD
101819-168
1,3-Dimethyl-5-iodopyrazole
1,3-Dimethyl-5-iodopyrazole

Hazard: Irritant

Formula: C₅H₇IN₂
MDL Number: MFCD09743720
CAS Number: 85779-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR