2-[3-(Ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid

Supplier: Matrix Scientific

024888-500MG
101819-152EA 184.62 USD
101819-152
2-[3-(Ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid
2-[3-(Ethoxycarbonyl)piperidin-1-yl]-5-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₆
MDL Number: MFCD09064901
CAS Number: 1000018-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR