5-Bromomethyl-2-methyl-4-trifluoromethyl-1,3-thiazole

Supplier: Matrix Scientific

024885-250MG
101819-148EA 320 USD
101819-148
5-Bromomethyl-2-methyl-4-trifluoromethyl-1,3-thiazole
5-Bromomethyl-2-methyl-4-trifluoromethyl-1,3-thiazole

Hazard: Irritant

Formula: C₆H₅BrF₃NS
MW: 260.08 g/mol
MDL Number: MFCD09264554
CAS Number: 1000339-73-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR