2-Fluoro-4-methylsulfonylphenol

Supplier: Matrix Scientific

024879-500MG
101819-136EA 40 USD
101819-136
2-Fluoro-4-methylsulfonylphenol
2-Fluoro-4-methylsulfonylphenol

Hazard: Irritant

Formula: C₇H₇FO₃S
MW: 190.19 g/mol
MDL Number: MFCD09264549
CAS Number: 398456-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR