2-Fluoro-5-methylsulfonyl-3-nitrotoluene

Supplier: Matrix Scientific

024866-500MG
101819-108EA 310.77 USD
101819-108
2-Fluoro-5-methylsulfonyl-3-nitrotoluene
2-Fluoro-5-methylsulfonyl-3-nitrotoluene

Hazard: Irritant

Formula: C₈H₈FNO₄S
MW: 233.22 g/mol
MDL Number: MFCD09264540
CAS Number: 1000339-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR