4-(Methylsulfonyl)-3-nitrobenzotrifluoride

Supplier: Matrix Scientific

024864-500MG
101819-106EA 213.85 USD
101819-106
4-(Methylsulfonyl)-3-nitrobenzotrifluoride
4-(Methylsulphonyl)-3-nitrobenzotrifluoride

Hazard: Irritant

Formula: C₈H₆F₃NO₄S
MDL Number: MFCD00107523
CAS Number: 364-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR