4-Chloroindole-2-carbohydrazide

Supplier: Matrix Scientific

024858-250MG
101819-094EA 335.38 USD
101819-094
4-Chloroindole-2-carbohydrazide
4-Chloroindole-2-carbohydrazide

Hazard: Irritant

Formula: C₉H₈ClN₃O
MDL Number: MFCD09839312
CAS Number: 64932-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR