1-[(4-Methylsulfonyl)phenyl]piperidine-2-carboxylic acid

Supplier: Matrix Scientific

024857-500MG
101819-092EA 469.23 USD
101819-092
1-[(4-Methylsulfonyl)phenyl]piperidine-2-carboxylic acid
1-[(4-Methylsulfonyl)phenyl]piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄S
MDL Number: MFCD09800753
CAS Number: 1190092-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR