1-[4-(Ethylsulfonyl)-2-fluorophenyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024856-250MG
101819-090EA 300 USD
101819-090
1-[4-(Ethylsulfonyl)-2-fluorophenyl]piperidine-3-carboxylic acid
1-[4-(Ethylsulfonyl)-2-fluorophenyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈FNO₄S
MDL Number: MFCD09835168
CAS Number: 1020722-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR