1-(2-Cyanophenyl)piperidine-2-carboxylic acid

Supplier: Matrix Scientific

024854-250MG
101819-086EA 450.77 USD
101819-086
1-(2-Cyanophenyl)piperidine-2-carboxylic acid
1-(2-Cyanophenyl)piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂
MDL Number: MFCD09835166
CAS Number: 1103191-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR