3-(2-Methyl-4-nitroimidazol-1-yl)propionitrile ≥97%
Supplier: Matrix Scientific
024841-5G
024841-1G
101819-042EA
40
USD
101819-042
101819-044
3-(2-Methyl-4-nitroimidazol-1-yl)propionitrile ≥97%
3-(2-Methyl-4-nitroimidazol-1-yl)propionitrile
Hazard: Irritant
Formula:
C₇H₈N₄O₂ |
MDL Number:
MFCD00134458 CAS Number: 89128-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...