3-Fluoro-4-methoxybenzenesulfonamide ≥98%

Supplier: Matrix Scientific

024839-1G
101819-036EA 230.77 USD
101819-036
3-Fluoro-4-methoxybenzenesulfonamide ≥98%
3-Fluoro-4-methoxybenzenesulphonamide

Hazard: Irritant

Formula: C₇H₈FNO₃S
MW: 205.21
MDL Number: MFCD02091381
CAS Number: 874767-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR