2-Methoxy-4-(trifluoromethoxy)benzaldehyde ≥98%

Supplier: Matrix Scientific

024805-1G
101818-936EA 100 USD
101818-936
2-Methoxy-4-(trifluoromethoxy)benzaldehyde ≥98%
2-Methoxy-4-(trifluoromethoxy)benzaldehyde

Hazard: Irritant

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
MDL Number: MFCD04115965
CAS Number: 886500-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR