3-Iodo-7-nitroindazole ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Iodo-7-nitro-1H-indazole

024801-250MG
101818-928EA 449.23 USD
101818-928
3-Iodo-7-nitroindazole ≥98%
3-Iodo-7-nitroindazole

Hazard: Irritant

Formula: C₇H₄IN₃O₂
MW: 289.03 g/mol
MDL Number: MFCD09027004
CAS Number: 864724-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR