Diethyl [2-(indol-3-yl)-2-oxoethyl]phosphonate ≥97%

Supplier: Matrix Scientific

024789-1G
101818-906EA 415.38 USD
101818-906
Diethyl [2-(indol-3-yl)-2-oxoethyl]phosphonate ≥97%
Diethyl [2-(indol-3-yl)-2-oxoethyl]phosphonate

Hazard: Irritant

Formula: C₁₄H₁₈NO₄P
MDL Number: MFCD09839792
CAS Number: 850231-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR