3-(5-Bromoindol-3-yl)-3-oxopropanenitrile ≥97%

Supplier: Matrix Scientific

024787-1G
101818-902EA 476.92 USD
101818-902
3-(5-Bromoindol-3-yl)-3-oxopropanenitrile ≥97%
3-(5-Bromoindol-3-yl)-3-oxopropanenitrile

Hazard: Irritant

Formula: C₁₁H₇BrN₂O
MDL Number: MFCD09839790
CAS Number: 1020722-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR