5,11-Dihydroindolo[3,2-b]carbazole ≥97%

Supplier: Matrix Scientific

Synonyms: Indolo[3,2-b]carbazole

024785-10MG
101818-898EA 732.31 USD
101818-898
5,11-Dihydroindolo[3,2-b]carbazole ≥97%
5,11-Dihydroindolo[3,2-b]carbazole
Formula: C₁₈H₁₂N₂
MW: 256.31 g/mol
MDL Number: MFCD09879258
CAS Number: 6336-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR