2-Benzo[1,3]dioxol-5-ylpiperazine ≥95%

Supplier: Matrix Scientific

024777-250MG
101818-880EA 200 USD
101818-880
2-Benzo[1,3]dioxol-5-ylpiperazine ≥95%
2-Benzo[1,3]dioxol-5-ylpiperazine

Hazard: Irritant

Formula: C₁₁H₁₄N₂O₂
MW: 206.25 g/mol
MDL Number: MFCD06245451
CAS Number: 65709-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR