1-(2-Nitrophenyl)-3-phenylpiperazine ≥95%

Supplier: Matrix Scientific

024767-250MG
101818-858EA 315.38 USD
101818-858
1-(2-Nitrophenyl)-3-phenylpiperazine ≥95%
1-(2-Nitrophenyl)-3-phenylpiperazine

Hazard: Irritant

Formula: C₁₆H₁₇N₃O₂
MDL Number: MFCD07371418
CAS Number: 904818-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR