1-(3-Methylpyridin-2-yl)-1,4-diazepane ≥95%

Supplier: Matrix Scientific

024764-250MG
101818-850EA 243.08 USD
101818-850
1-(3-Methylpyridin-2-yl)-1,4-diazepane ≥95%
1-(3-Methylpyridin-2-yl)-1,4-diazepane

Hazard: Irritant

Formula: C₁₁H₁₇N₃
MDL Number: MFCD08061111
CAS Number: 880362-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR