2-(2-Methoxyphenyl)piperazine ≥95%

Supplier: Matrix Scientific

024755-250MG
101818-828EA 386.15 USD
101818-828
2-(2-Methoxyphenyl)piperazine ≥95%
2-(2-Methoxyphenyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD06660448
CAS Number: 65709-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR