Dimethyl(4-piperazin-2-ylphenyl)amine ≥95%

Supplier: Matrix Scientific

024749-250MG
101818-810EA 455.38 USD
101818-810
Dimethyl(4-piperazin-2-ylphenyl)amine ≥95%
Dimethyl(4-piperazin-2-ylphenyl)amine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MDL Number: MFCD06660429
CAS Number: 904814-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR