5-Bromo-2-hydroxy-4-picoline ≥98%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-hydroxy-4-methylpyridine
024734-5G
024734-25G
101818-770EA
200
USD
101818-770
101818-768
5-Bromo-2-hydroxy-4-picoline ≥98%
5-Bromo-2-hydroxy-4-picoline
Hazard: Irritant
Formula:
C₆H₆BrNO MW: 188.02 g/mol |
MDL Number:
MFCD03427664 CAS Number: 164513-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...