2-Mercapto-6-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Sulphanyl-6-(trifluoromethyl)pyridine

024711-1G
101818-710EA 101.54 USD
101818-710
2-Mercapto-6-(trifluoromethyl)pyridine ≥98%
2-Mercapto-6-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₄F₃NS
MW: 179.16 g/mol
MDL Number: MFCD09864680
CAS Number: 121307-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR