5-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥97%

Supplier: Matrix Scientific

024685-250MG
101818-638EA 444.62 USD
101818-638
5-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine ≥97%
5-Fluoro-3-iodo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₄FIN₂
MDL Number: MFCD08688599
CAS Number: 900514-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR