3-Bromo-2,6-dimethylpyridine ≥98%

Supplier: Matrix Scientific

024666-5G
101818-592EA 46.15 USD
101818-592
3-Bromo-2,6-dimethylpyridine ≥98%
3-Bromo-2,6-dimethylpyridine

Hazard: Irritant

Formula: C₇H₈BrN
MW: 186.0491 g/mol
Boiling Pt: 56…58 °C
MDL Number: MFCD00235164
CAS Number: 3430-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR