5-Acetyl-4-methyl-2-(2-pyrazinyl)thiazole

Supplier: Matrix Scientific

024582-500MG
101818-364EA 280 USD
101818-364
5-Acetyl-4-methyl-2-(2-pyrazinyl)thiazole
5-Acetyl-4-methyl-2-(2-pyrazinyl)thiazole

Hazard: Irritant

Formula: C₁₀H₉N₃OS
MDL Number: MFCD01764839
CAS Number: 260789-23-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR