2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone

Supplier: Matrix Scientific

Synonyms: 4-(Bromoacetyl)-5-methyl-1-phenyl-1H-pyrazole

024580-500MG
101818-360EA 238.46 USD
101818-360
2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone
2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone

Hazard: Irritant

Formula: C₁₂H₁₁BrN₂O
MW: 279.14 g/mol
MDL Number: MFCD02681921
CAS Number: 137577-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR