Ethyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Supplier: Matrix Scientific

024579-500MG
101818-358EA 253.85 USD
101818-358
Ethyl-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate
Ethyl 2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₉F₃N₂O₂
MDL Number: MFCD06496237
CAS Number: 108438-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR