5-Bromobenzo[b]furan-2-carbaldehyde

Supplier: Matrix Scientific

024570-500MG
101818-340EA 204.62 USD
101818-340
5-Bromobenzo[b]furan-2-carbaldehyde
5-Bromobenzo[b]furan-2-carbaldehyde

Hazard: Irritant

Formula: C₉H₅BrO₂
MW: 225.05 g/mol
MDL Number: MFCD06659072
CAS Number: 23145-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR