1-[(2-Methyl-4-methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024564-250MG
101818-328EA 235.38 USD
101818-328
1-[(2-Methyl-4-methylsulfonyl)phenyl]piperidine-3-carboxylic acid
1-[(2-Methyl-4-methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₉NO₄S
MDL Number: MFCD09038320
CAS Number: 942474-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR