2,6-Dimethyl-4-hydrazinopyrimidine

Supplier: Matrix Scientific

024558-500MG
101818-318EA 140 USD
101818-318
2,6-Dimethyl-4-hydrazinopyrimidine
2,6-Dimethyl-4-hydrazinopyrimidine

Hazard: Irritant

Formula: C₆H₁₀N₄
MDL Number: MFCD00102150
CAS Number: 14331-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR