5-Methyl-1-phenylpyrazole-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 5-Methyl-1-phenyl-1H-pyrazole-3-carboxylic acid

024553-500MG
101818-308EA 306.15 USD
101818-308
5-Methyl-1-phenylpyrazole-3-carboxylic acid
5-Methyl-1-phenylpyrazole-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MW: 202.21 g/mol
MDL Number: MFCD00124910
CAS Number: 10199-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR