Ethyl-3-methyl-1-phenylpyrazole-5-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 3-methyl-1-phenyl-1H-pyrazole-5-carboxylate

024552-250MG
101818-306EA 78.46 USD
101818-306
Ethyl-3-methyl-1-phenylpyrazole-5-carboxylate
Ethyl-3-methyl-1-phenylpyrazole-5-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₂
MW: 230.27 g/mol
MDL Number: MFCD00221432
CAS Number: 81153-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR