1-[2-Chloro-5-methyl-4-(methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024547-250MG
101818-294EA 224.62 USD
101818-294
1-[2-Chloro-5-methyl-4-(methylsulfonyl)phenyl]piperidine-3-carboxylic acid
1-[2-Chloro-5-methyl-4-(methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₈ClNO₄S
MDL Number: MFCD09027742
CAS Number: 942474-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR