1-(4-Amino-2-nitrophenyl)piperidine-4-carboxamide

Supplier: Matrix Scientific

024543-500MG
101818-288EA 186.15 USD
101818-288
1-(4-Amino-2-nitrophenyl)piperidine-4-carboxamide
1-(4-Amino-2-nitrophenyl)piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₆N₄O₃
MDL Number: MFCD09027738
CAS Number: 942474-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR