2,6-Dimethyl-4-[2-nitro-4-(trifluoromethyl)phenyl]morpholine

Supplier: Matrix Scientific

024532-1G
101818-268EA 170.77 USD
101818-268
2,6-Dimethyl-4-[2-nitro-4-(trifluoromethyl)phenyl]morpholine
2,6-Dimethyl-4-[2-nitro-4-(trifluoromethyl)phenyl]morpholine

Hazard: Irritant

Formula: C₁₃H₁₅F₃N₂O₃
MDL Number: MFCD02018696
CAS Number: 405910-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR