1-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine-2-carboxylic acid
Supplier: Matrix Scientific
024531-500MG
101818-266EA
186.15
USD
101818-266
1-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine-2-carboxylic acid
1-[2-Nitro-4-(trifluoromethyl)phenyl]pyrrolidine-2-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₁F₃N₂O₄ |
MDL Number:
MFCD01420274 CAS Number: 1008859-90-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...