1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine

Supplier: Matrix Scientific

Synonyms: 4-Methyl-1-[2-nitro-4-(trifluoromethyl)phenyl]piperidine

024528-1G
101818-260EA 170.77 USD
101818-260
1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine
1-[2-Nitro-4-(trifluoromethyl)phenyl]-4-methylpiperidine

Hazard: Irritant

Formula: C₁₃H₁₅F₃N₂O₂
MW: 288.27 g/mol
MDL Number: MFCD09027734
CAS Number: 392237-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR