1-Methyl-4-[2-nitro-4-(trifluoromethyl)phenyl]piperazine hydrochloride

Supplier: Matrix Scientific

024526-1G
101818-256EA 100 USD
101818-256
1-Methyl-4-[2-nitro-4-(trifluoromethyl)phenyl]piperazine hydrochloride
1-Methyl-4-[2-nitro-4-(trifluoromethyl)phenyl]piperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃N₃O₂
MDL Number: MFCD01208354
CAS Number: 298703-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR