1-[(2-Chloro-4-methylsulfonyl)phenyl]piperidine-3-carbohydrazide

Supplier: Matrix Scientific

024524-250MG
101818-252EA 272.31 USD
101818-252
1-[(2-Chloro-4-methylsulfonyl)phenyl]piperidine-3-carbohydrazide
1-[(2-Chloro-4-methylsulfonyl)phenyl]piperidine-3-carbohydrazide

Hazard: Irritant

Formula: C₁₃H₁₈ClN₃O₃S
MDL Number: MFCD09038314
CAS Number: 951625-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR