4-[3-(Ethoxycarbonyl)piperidin-1-yl]-3-nitrobenzoic acid

Supplier: Matrix Scientific

024523-500MG
101818-250EA 243.08 USD
101818-250
4-[3-(Ethoxycarbonyl)piperidin-1-yl]-3-nitrobenzoic acid
4-[3-(Ethoxycarbonyl)piperidin-1-yl]-3-nitrobenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₈N₂O₆
MDL Number: MFCD09027733
CAS Number: 942474-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR