Ethyl-3-nitro-4-(azepan-1-yl)benzoate

Supplier: Matrix Scientific

024514-1G
101818-234EA 267.69 USD
101818-234
Ethyl-3-nitro-4-(azepan-1-yl)benzoate
Ethyl-3-nitro-4-(azepan-1-yl)benzoate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₄
MDL Number: MFCD09038311
CAS Number: 71302-98-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR