3-Nitro-4-(piperidin-4-carboxamide-1-yl)benzoic acid

Supplier: Matrix Scientific

024507-500MG
101818-220EA 186.15 USD
101818-220
3-Nitro-4-(piperidin-4-carboxamide-1-yl)benzoic acid
3-Nitro-4-(piperidin-4-carboxamide-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₅
MDL Number: MFCD02937553
CAS Number: 440347-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR