5-Nitro-2-(piperidin-4-carboxamide-1-yl)benzoic acid

Supplier: Matrix Scientific

024505-500MG
101818-216EA 186.15 USD
101818-216
5-Nitro-2-(piperidin-4-carboxamide-1-yl)benzoic acid
5-Nitro-2-(piperidin-4-carboxamide-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₅
MW: 293.28 g/mol
MDL Number: MFCD09038305
CAS Number: 942474-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR