5-Nitro-2-(piperidin-4-ol-1-yl)benzoic acid

Supplier: Matrix Scientific

024502-1G
101818-210EA 267.69 USD
101818-210
5-Nitro-2-(piperidin-4-ol-1-yl)benzoic acid
5-Nitro-2-(piperidin-4-ol-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₅
MDL Number: MFCD09038304
CAS Number: 942474-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR