Pyrazine-2-carboximidic acid methyl ester hydrochloride

Supplier: Matrix Scientific

024495-500MG
101818-196EA 307.69 USD
101818-196
Pyrazine-2-carboximidic acid methyl ester hydrochloride
Pyrazine-2-carboximidic acid methyl ester hydrochloride

Hazard: Irritant

Formula: C₆H₈ClN₃O
MDL Number: MFCD10697992
CAS Number: 74617-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR