5-(3-Methylpiperidin-1-yl)-2-methylsulfonylaniline

Supplier: Matrix Scientific

024494-500MG
101818-194EA 267.69 USD
101818-194
5-(3-Methylpiperidin-1-yl)-2-methylsulfonylaniline
5-(3-Methylpiperidin-1-yl)-2-methylsulfonylaniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD09027724
CAS Number: 942474-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR