4'-(Ethylsulfonyl)acetophenone

024493-500MG
101818-192EA 196.65 USD
101818-192
4'-(Ethylsulfonyl)acetophenone
4'-(Ethylsulfonyl)acetophenone

Hazard: Irritant

Formula: C₁₀H₁₂O₃S
CAS Number: 99186-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR